Gene Transcripts

Gene Symbol
SLC5A4
Entrez Gene ID
6527
Gene Name
solute carrier family 5 member 4
Gene Aliases
DJ90G24.4, SAAT1, SGLT3
Chromosome Location
Chr. 22: 32218423 - 32355057 on Build GRCh38
UniGene ID
Hs.130101
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014227.2 NP_055042.1 8 692
XM_006724308.3 XP_006724371.1 9 943
XM_011530342.2 XP_011528644.1 10 1061
XM_011530343.2 XP_011528645.1 8 607
XM_011530344.2 XP_011528646.1 7 572
XM_017028920.1 XP_016884409.1 10 1022
GenBank mRNA AJ133127.1 - 8 706
U41897.1 - 6 475

View Full Product Details