Gene Transcripts

Gene Symbol
PRKCI
Entrez Gene ID
5584
Gene Name
protein kinase C iota
Gene Aliases
DXS1179E, PKCI, nPKC-iota
Chromosome Location
Chr. 3: 170222432 - 170305982 on Build GRCh38
UniGene ID
Hs.478199
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002740.5 NP_002731.4 9 965
XM_017006860.1 XP_016862349.1 8 872
GenBank mRNA AB451443.1 - 9 700
AK315342.1 - 9 893
BC022016.2 - 9 960 4823853
L18964.1 - 9 964
L33881.1 - 9 904

View Full Product Details