Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001282161.1 | NP_001269090.1 | 6 | 1423 | |
NM_003336.3 | NP_003327.2 | 6 | 1430 | ||
NM_181762.2 | NP_861427.1 | 5 | 1340 | ||
GenBank mRNA | AK223045.1 | - | 6 | 1332 | |
AK223064.1 | - | 6 | 1329 | ||
AK297696.1 | - | 5 | 1242 | ||
BC042021.1 | - | 3 | 2207 | ||
M74524.1 | - | 6 | 1305 |