Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003883.3 | NP_003874.2 | 9 | 800 | |
XM_011537697.2 | XP_011535999.1 | 8 | 652 | ||
GenBank mRNA | AF005482.1 | - | 7 | 816 | |
AF039703.1 | - | 9 | 756 | ||
BC000614.1 | - | 9 | 789 | 3345755 | |
U66914.1 | - | 9 | 745 | ||
U75696.1 | - | 8 | 783 | ||
U75697.1 | - | 9 | 789 |