Gene Transcripts

Gene Symbol
OPTC
Entrez Gene ID
26254
Gene Name
opticin
Gene Aliases
OPT
Chromosome Location
Chr. 1: 203494143 - 203508949 on Build GRCh38
UniGene ID
Hs.632468
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014359.3 NP_055174.1 4, 5 647
XM_011509406.2 XP_011507708.1 3, 4 1046
GenBank mRNA AF161702.1 - 4, 5 624
AJ133790.2 - 4, 5 647
AY077681.1 - 4, 5 647
BC074942.2 - 3, 4 565 30915495
BC074943.2 - 3, 4 565 30915272

View Full Product Details