Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001130012.2 | NP_001123484.1 | 7 | 1545 | |
NM_001252073.1 | NP_001239002.1 | 6 | 1162 | ||
NM_001252075.1 | NP_001239004.1 | 6 | 1156 | ||
NM_001252076.1 | NP_001239005.1 | 6 | 1129 | ||
NM_004785.5 | NP_004776.3 | 7 | 1512 | ||
XM_005255702.4 | XP_005255759.1 | 6 | 2225 | ||
XM_006720981.1 | XP_006721044.1 | 6 | 3022 | ||
XM_017023894.1 | XP_016879383.1 | 6 | 2115 | ||
XM_017023895.1 | XP_016879384.1 | 7 | 1400 | ||
GenBank mRNA | AF004900.1 | - | 7 | 1423 | |
BC014513.1 | - | 7 | 1518 | ||
BC069014.1 | - | 7 | 1514 | ||
BC106001.1 | - | 7 | 1521 | 5296143 | |
U82108.1 | - | 7 | 1465 | ||
Z50150.1 | - | 7 | 1499 |