Gene Transcripts

Gene Symbol
OR51F2
Entrez Gene ID
119694
Gene Name
olfactory receptor family 51 subfamily F member 2
Gene Aliases
OR11-23
Chromosome Location
Chr. 11: 4821386 - 4822414 on Build GRCh38
UniGene ID
Hs.553562
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004753.1 NP_001004753.1 1 980
GenBank mRNA BC136877.1 - 1 1045

View Full Product Details