Gene Transcripts

Gene Symbol
KRTAP13-4
Entrez Gene ID
284827
Gene Name
keratin associated protein 13-4
Gene Aliases
KAP13.4
Chromosome Location
Chr. 21: 30430276 - 30430758 on Build GRCh38
UniGene ID
Hs.553677
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181600.1 NP_853631.1 1 355
GenBank mRNA AB096941.1 - 1 377
BC131691.1 - 1 359 40119689

View Full Product Details