Gene Transcripts
- Gene Symbol
- OR4N2
- Entrez Gene ID
- 390429
- Gene Name
- olfactory receptor family 4 subfamily N member 2
- Gene Aliases
- OR14-13, OR14-8
- Chromosome Location
- Chr. 14: 19803770 - 19828372 on Build GRCh38
- UniGene ID
- Hs.512490
- Species
- Homo sapiens
- Gene Symbol
- OR4N3P
- Entrez Gene ID
- 390539
- Gene Name
- olfactory receptor family 4 subfamily N member 3 pseudogene
- Gene Aliases
- OR15-2
- Chromosome Location
- Chr. 15: 22125511 - 22126447 on Build GRCh38
- UniGene ID
- Hs.666555
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028067.1 |
|
1 |
27 |
|
View Full Product Details