Gene Transcripts

Gene Symbol
OR2B3
Entrez Gene ID
442184
Gene Name
olfactory receptor family 2 subfamily B member 3
Gene Aliases
6M1-1, OR2B3P, OR6-14, OR6-4
Chromosome Location
Chr. 6: 29086208 - 29087313 on Build GRCh38
UniGene ID
Hs.553835
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005226.2 NP_001005226.1 1 487
GenBank mRNA BC136817.1 - 1 446
BC136818.1 - 1 446

View Full Product Details