Gene Transcripts

Gene Symbol
OR5M8
Entrez Gene ID
219484
Gene Name
olfactory receptor family 5 subfamily M member 8
Gene Aliases
OR11-194
Chromosome Location
Chr. 11: 56490435 - 56491370 on Build GRCh38
UniGene ID
Hs.554522
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005282.1 NP_001005282.1 1 903
GenBank mRNA BC136978.1 - 1 928
BC136980.1 - 1 928 9020972

View Full Product Details