Gene Transcripts

Gene Symbol
CFHR2
Entrez Gene ID
3080
Gene Name
complement factor H related 2
Gene Aliases
CFHL2, FHR2, HFL3
Chromosome Location
Chr. 1: 196943752 - 196959328 on Build GRCh38
UniGene ID
Hs.632450
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005666.3 NP_005657.1 1, 2 179
XM_011509458.2 XP_011507760.1 1, 2 163
XM_017001109.1 XP_016856598.1 1, 2 163
GenBank mRNA BC022283.1 - 1, 2 168
BG563114.1 - 1, 2 180
X64877.1 - 1, 2 143

View Full Product Details