Gene Transcripts

Gene Symbol
RRAGD
Entrez Gene ID
58528
Gene Name
Ras related GTP binding D
Gene Aliases
RAGD, bA11D8.2.1
Chromosome Location
Chr. 6: 89363509 - 89412470 on Build GRCh38
UniGene ID
Hs.31712
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021244.4 NP_067067.1 6 1240
XM_011536022.2 XP_011534324.1 6 1012
GenBank mRNA AF272036.1 - 6 1023
AK289799.1 - 6 1165
AL137502.1 - 5 738
BC003088.1 - 6 902 3536088

View Full Product Details