Gene Transcripts

Gene Symbol
LRRIQ1
Entrez Gene ID
84125
Gene Name
leucine rich repeats and IQ motif containing 1
Gene Aliases
Chromosome Location
Chr. 12: 85036321 - 85273004 on Build GRCh38
UniGene ID
Hs.402200
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001079910.1 NP_001073379.1 8 1590
XM_011538817.2 XP_011537119.1 8 1533
XM_011538818.2 XP_011537120.1 8 1533
XM_011538819.2 XP_011537121.1 8 1533
XM_011538823.2 XP_011537125.1 8 1533
XM_011538824.2 XP_011537126.1 8 1533
XM_011538825.2 XP_011537127.1 8 1533
XM_017020019.1 XP_016875508.1 8 1533
GenBank mRNA AB058704.1 - 4 1154
AK022365.1 - 8 1591
AK307799.1 - 8 1541

View Full Product Details