Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001319238.1 | NP_001306167.1 | 20 | 4854 | |
NM_001319239.1 | NP_001306168.1 | 20 | 4749 | ||
NM_014827.5 | NP_055642.3 | 20 | 4425 | ||
XM_005245643.2 | XP_005245700.1 | 20 | 4444 | ||
XM_005245644.3 | XP_005245701.1 | 19 | 4128 | ||
XM_006711673.2 | XP_006711736.1 | 20 | 4291 | ||
XM_011510199.1 | XP_011508501.1 | 18 | 5496 | ||
XM_011510200.1 | XP_011508502.1 | 19 | 4516 | ||
XM_011510201.1 | XP_011508503.1 | 20 | 4702 | ||
XM_011510202.1 | XP_011508504.1 | 18 | 4441 | ||
XM_017002956.1 | XP_016858445.1 | 19 | 5750 | ||
XM_017002957.1 | XP_016858446.1 | 19 | 5602 | ||
XM_017002958.1 | XP_016858447.1 | 19 | 5589 | ||
XM_017002959.1 | XP_016858448.1 | 20 | 4662 | ||
XM_017002960.1 | XP_016858449.1 | 19 | 5744 | ||
XM_017002961.1 | XP_016858450.1 | 18 | 5490 | ||
XM_017002962.1 | XP_016858451.1 | 19 | 4510 | ||
XM_017002963.1 | XP_016858452.1 | 20 | 4302 | ||
XM_017002964.1 | XP_016858453.1 | 19 | 5662 | ||
XM_017002965.1 | XP_016858454.1 | 18 | 4985 | ||
GenBank mRNA | AB014563.1 | - | 17 | 3936 | |
AL049419.1 | - | 1 | 1122 | ||
BC038513.2 | - | 20 | 4580 | ||
BX649148.1 | - | 17 | 8061 | ||
CR627439.1 | - | 19 | 5446 | ||
CR627446.1 | - | 17 | 7549 |