Gene Transcripts

Gene Symbol
MIR1-1HG
Entrez Gene ID
128826
Gene Name
MIR1-1 host gene
Gene Aliases
C20orf166, MIR133A2HG, dJ353C17.1
Chromosome Location
Chr. 20: 62550453 - 62570764 on Build GRCh38
UniGene ID
Hs.86507
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001302812.1 NP_001289741.1 5 808
NM_178463.3 NP_848558.1 4 1189
GenBank mRNA AK056985.1 - 4 1205
F35210.1 - 1 161

View Full Product Details