Gene Transcripts

Gene Symbol
mrpl11
Entrez Gene ID
293666
Gene Name
mitochondrial ribosomal protein L11
Gene Aliases
Chromosome Location
Chr. 1: 220243471 - 220246340 on Build Rnor_6.0
UniGene ID
Rn.3762
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001006973.1 NP_001006974.1 3 301
XM_006230739.2 XP_006230801.1 3 345
GenBank mRNA BC083740.1 - 3 301
FQ225172.1 - 3 326

View Full Product Details