Gene Transcripts
- Gene Symbol
- Rhno1
- Entrez Gene ID
- 297627
- Gene Name
- RAD9-HUS1-RAD1 interacting nuclear orphan 1
- Gene Aliases
- Chromosome Location
- Chr. 4: 161679674 - 161685060 on Build Rnor_6.0
- UniGene ID
- Rn.1747
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC079219.1 |
- |
3 |
1433 |
|
- Gene Symbol
- LOC100362315
- Entrez Gene ID
- 100362315
- Gene Name
- Uncharacterized protein C12orf32 homolog
- Gene Aliases
- Chromosome Location
- Chr. 2: 199263118 - 199264218 on Build Rnor_6.0
- Species
- Rattus norvegicus
View Full Product Details