Gene Transcripts

Gene Symbol
Tor2a
Entrez Gene ID
362112
Gene Name
torsin family 2, member A
Gene Aliases
Chromosome Location
Chr. 3: 11794727 - 11800261 on Build Rnor_6.0
UniGene ID
Rn.13187
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007744.1 NP_001007745.1 3, 4 651
XM_006233981.2 XP_006234043.1 3, 4 1286
GenBank mRNA BC078943.1 - 3, 4 651

View Full Product Details