Gene Transcripts

Gene Symbol
PIWIL3
Entrez Gene ID
440822
Gene Name
piwi like RNA-mediated gene silencing 3
Gene Aliases
HIWI3
Chromosome Location
Chr. 22: 24719034 - 24774720 on Build GRCh38
UniGene ID
Hs.448343
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008496.3 NP_001008496.2 21 3093
NM_001255975.1 NP_001242904.1 21 3066
NR_045648.1 22 3276
NR_045649.1 22 3149
GenBank mRNA AB079368.1 - 21 3089
AK302409.1 - 21 3066
BC144693.1 - 22 2906
BC144696.1 - 22 3033

View Full Product Details