Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001008884.2 | NP_001008884.1 | 3 | 652 | |
GenBank mRNA | AF084933.1 | - | 3 | 592 | |
AY626207.1 | - | 3 | 588 | ||
BC087691.1 | - | 4 | 572 | 7310949 | |
CO563535.1 | - | 3 | 652 | ||
FQ225193.1 | - | 3 | 640 | ||
FQ226724.1 | - | 3 | 640 | ||
FQ226759.1 | - | 3 | 649 | ||
FQ226850.1 | - | 3 | 640 | ||
FQ227087.1 | - | 3 | 637 | ||
FQ228005.1 | - | 3 | 640 | ||
FQ228124.1 | - | 3 | 617 | ||
FQ228129.1 | - | 3 | 640 | ||
FQ228185.1 | - | 3 | 658 | ||
FQ230360.1 | - | 3 | 640 | ||
FQ231387.1 | - | 3 | 640 | ||
FQ231709.1 | - | 3 | 640 | ||
FQ232046.1 | - | 3 | 640 | ||
FQ232100.1 | - | 3 | 639 | ||
FQ232453.1 | - | 3 | 640 | ||
FQ233049.1 | - | 3 | 636 | ||
FQ233335.1 | - | 3 | 640 | ||
FQ233396.1 | - | 3 | 640 | ||
FQ233493.1 | - | 3 | 640 | ||
FQ233800.1 | - | 3 | 618 | ||
FQ234464.1 | - | 3 | 639 | ||
FQ234907.1 | - | 3 | 640 | ||
KC222950.1 | - | 3 | 588 | ||
X53054.1 | - | 3 | 603 |