Gene Transcripts

Gene Symbol
Dnal1
Entrez Gene ID
105000
Gene Name
dynein, axonemal, light chain 1
Gene Aliases
1700010H15Rik, AW121714, Dnalc1, E330027P08Rik
Chromosome Location
Chr. 12: 84114280 - 84143517 on Build GRCm38
UniGene ID
Mm.101525
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028821.3 NP_083097.2 3 200
GenBank mRNA AK005826.1 - 3 286
BC070454.1 - 157
BC125392.1 - 2 182 40129907
BC125394.1 - 2 178 40129909

View Full Product Details