Gene Transcripts

Gene Symbol
Mrgprx2l
Entrez Gene ID
404645
Gene Name
MAS-related G protein-coupled receptor, member X2-like
Gene Aliases
MrgB2, Mrgprb2
Chromosome Location
Chr. 1: 103810204 - 103811148 on Build Rnor_6.0
UniGene ID
Rn.141966
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002285.1 NP_001002285.1 1 934

View Full Product Details