Gene Transcripts

Gene Symbol
Pnkp
Entrez Gene ID
308576
Gene Name
polynucleotide kinase 3'-phosphatase
Gene Aliases
Chromosome Location
Chr. 1: 100853474 - 100859202 on Build Rnor_6.0
UniGene ID
Rn.45061
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004259.1 NP_001004259.1 1, 2 220
XM_006229058.2 XP_006229120.1 1, 2 652
GenBank mRNA BC079307.1 - 1, 2 220

View Full Product Details