Gene Transcripts

Gene Symbol
Tbc1d20
Entrez Gene ID
362237
Gene Name
TBC1 domain family, member 20
Gene Aliases
Chromosome Location
Chr. 3: 147772006 - 147790946 on Build Rnor_6.0
UniGene ID
Rn.206296
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004281.1 NP_001004281.1 6 760
GenBank mRNA BC079383.1 - 6 760

View Full Product Details