Gene Transcripts

Gene Symbol
Zmynd10
Entrez Gene ID
363139
Gene Name
zinc finger, MYND-type containing 10
Gene Aliases
Chromosome Location
Chr. 8: 116302513 - 116306871 on Build Rnor_6.0
UniGene ID
Rn.75726
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004284.1 NP_001004284.1 5 509
GenBank mRNA BC079255.1 - 5 509

View Full Product Details