Gene Transcripts

Gene Symbol
ANKRD34B
Entrez Gene ID
340120
Gene Name
ankyrin repeat domain 34B
Gene Aliases
DP58
Chromosome Location
Chr. 5: 80556755 - 80570485 on Build GRCh38
UniGene ID
Hs.519404
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004441.2 NP_001004441.2 5 3046
XM_006714599.3 XP_006714662.1 5 2808
XM_011543366.2 XP_011541668.1 5 2850
XM_011543369.2 XP_011541671.1 3 2806
GenBank mRNA BC142957.1 - 4 2726
CR749535.1 - 5 2840

View Full Product Details