Gene Transcripts

Gene Symbol
Txlna
Entrez Gene ID
109658
Gene Name
taxilin alpha
Gene Aliases
2600010N21Rik, AI425952, AW209241, Il14, Txln
Chromosome Location
Chr. 4: 129626077 - 129641084 on Build GRCm38
UniGene ID
Mm.268863
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005506.3 NP_001005506.2 10 4805
NM_001199695.1 NP_001186624.1 11 4522
GenBank mRNA AK011182.1 - 1 651
AK150240.1 - 11 4479
BC060227.1 - 10 4640 30357541
BC065115.1 - 9 4876 30362350

View Full Product Details