Gene Transcripts

Gene Symbol
Itgb4
Entrez Gene ID
192897
Gene Name
integrin beta 4
Gene Aliases
AA407042, C230078O20, CD104
Chromosome Location
Chr. 11: 115974725 - 116008412 on Build GRCm38
UniGene ID
Mm.213873
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005608.2 NP_001005608.2 26 3520
NM_133663.2 NP_598424.2 26 3520
GenBank mRNA AK048888.1 - 26 3405
AK144111.1 - 3 516
AK145317.1 - 3 596
AK147424.1 - 26 3518
BC006632.1 - 6 566
BC059192.1 - 22 2715
BF139425.1 - 1 142
L04678.1 - 27 3364

View Full Product Details