Gene Transcripts

Gene Symbol
Dhx40
Entrez Gene ID
287595
Gene Name
DEAH (Asp-Glu-Ala-His) box polypeptide 40
Gene Aliases
Chromosome Location
Chr. 10: 74081442 - 74119032 on Build Rnor_6.0
UniGene ID
Rn.29182
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005873.1 NP_001005873.1 5 842
GenBank mRNA BC083821.1 - 5 842

View Full Product Details