Gene Transcripts

Gene Symbol
ZMIZ2
Entrez Gene ID
83637
Gene Name
zinc finger MIZ-type containing 2
Gene Aliases
NET27, TRAFIP20, ZIMP7, hZIMP7
Chromosome Location
Chr. 7: 44748547 - 44769881 on Build GRCh38
UniGene ID
Hs.600681
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001300959.1 NP_001287888.1 18 2674
NM_031449.3 NP_113637.3 19 2843
NM_174929.2 NP_777589.2 17 2704
XM_005249866.2 XP_005249923.1 19 2894
XM_005249867.4 XP_005249924.1 19 2896
XM_005249868.4 XP_005249925.1 19 3914
XM_005249869.2 XP_005249926.1 19 2867
XM_005249870.2 XP_005249927.1 18 2816
XM_005249871.2 XP_005249928.1 19 2798
XM_005249872.2 XP_005249929.1 18 2789
XM_005249873.2 XP_005249930.1 19 2771
XM_011515566.1 XP_011513868.1 13 2034
XM_017012674.1 XP_016868163.1 19 2975
GenBank mRNA AB015330.1 - 4 719
AB067473.1 - 13 1927
AF357908.1 - 18 2726
AK057115.1 - 4 1304
AK090415.1 - 19 2843
AK090841.1 - 11 2123
AK128359.1 - 1 725
AK301932.1 - 6 583
AL136572.1 - 9 1296
AL831933.1 - 10 2441
AY426594.1 - 17 2666
BC021924.1 - 13 1775
BC110435.1 - 18 2659 5518312
BC131780.1 - 4 380

View Full Product Details