Gene Transcripts

Gene Symbol
ZMIZ2
Entrez Gene ID
83637
Gene Name
zinc finger MIZ-type containing 2
Gene Aliases
NET27, TRAFIP20, ZIMP7, hZIMP7
Chromosome Location
Chr. 7: 44748547 - 44769881 on Build GRCh38
UniGene ID
Hs.600681
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001300959.1 NP_001287888.1 8 1168
NM_031449.3 NP_113637.3 9 1337
NM_174929.2 NP_777589.2 7 1198
XM_005249866.2 XP_005249923.1 9 1361
XM_005249867.4 XP_005249924.1 9 1363
XM_005249868.4 XP_005249925.1 9 2381
XM_005249869.2 XP_005249926.1 9 1361
XM_005249870.2 XP_005249927.1 8 1283
XM_005249871.2 XP_005249928.1 9 1265
XM_005249872.2 XP_005249929.1 8 1283
XM_005249873.2 XP_005249930.1 9 1265
XM_011515566.1 XP_011513868.1 3 501
XM_017012674.1 XP_016868163.1 9 1442
GenBank mRNA AB067473.1 - 3 421
AF357908.1 - 8 1214
AK090841.1 - 1 617
AL831933.1 - 1 834
AY426594.1 - 7 1160
BC016968.1 - 7 1181
BC021924.1 - 3 269
BC110435.1 - 8 1153 5518312

View Full Product Details