Gene Transcripts

Gene Symbol
PSMD6
Entrez Gene ID
9861
Gene Name
proteasome 26S subunit, non-ATPase 6
Gene Aliases
Rpn7, S10, SGA-113M, p42A, p44S10
Chromosome Location
Chr. 3: 64010549 - 64024010 on Build GRCh38
UniGene ID
Hs.152536
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271779.1 NP_001258708.1 4, 5 843
NM_001271780.1 NP_001258709.1 3, 4 558
NM_001271781.1 NP_001258710.1 3, 4 433
NM_014814.2 NP_055629.1 3, 4 684
XM_005265619.1 XP_005265676.1 3, 4 780
XM_011534288.1 XP_011532590.1 4, 5 739
XM_017007569.1 XP_016863058.1 4, 5 586
GenBank mRNA AF215935.1 - 3, 4 550
AF530062.1 - 3, 4 497
AK095059.1 - 2, 3 1266
AK290205.1 - 3, 4 538
AL529900.3 - 3, 4 419
AY359879.1 - 3, 4 530
AY568086.1 - 3, 4 522
BC000630.1 - 3, 4 514 3344973
BC000904.2 - 3, 4 535 2900971
BC012369.1 - 3, 4 532 3858094
BG723464.1 - 3, 4 561
BX647742.1 - 4, 5 710
D14663.1 - 3, 4 522

View Full Product Details