Gene Transcripts
- Gene Symbol
- ZC3H11A
- Entrez Gene ID
- 9877
- Gene Name
- zinc finger CCCH-type containing 11A
- Gene Aliases
- ZC3HDC11A
- Chromosome Location
- Chr. 1: 203795537 - 203854128 on Build GRCh38
- UniGene ID
- Hs.532399
- Species
- Homo sapiens
- Gene Symbol
- LOC441155
- Entrez Gene ID
- 441155
- Gene Name
- zinc finger CCCH-type domain-containing-like
- Gene Aliases
- Chromosome Location
- Chr. 6: 65301418 - 65306069 on Build GRCh38
- UniGene ID
- Hs.546686
- Species
- Homo sapiens
View Full Product Details