Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001320808.1 | NP_001307737.1 | 3 | 1137 | |
NM_014873.2 | NP_055688.1 | 3 | 548 | ||
XM_005273362.3 | XP_005273419.1 | 4 | 696 | ||
XM_011510228.2 | XP_011508530.1 | 4 | 585 | ||
XM_011510229.2 | XP_011508531.1 | 3 | 468 | ||
XM_017003004.1 | XP_016858493.1 | 2 | 1700 | ||
GenBank mRNA | AY561706.1 | - | 2 | 321 | |
BC034621.1 | - | 3 | 1138 | 4839400 | |
BP369253.1 | - | 3 | 432 | ||
D86960.1 | - | 3 | 548 |