Gene Transcripts

Gene Symbol
Lcn6
Entrez Gene ID
414138
Gene Name
lipocalin 6
Gene Aliases
Chromosome Location
Chr. 3: 2888103 - 2893498 on Build Rnor_6.0
UniGene ID
Rn.124523
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001519.1 NP_001001519.1 2 194
GenBank mRNA AF303086.1 - 2 194

View Full Product Details