Gene Transcripts

Gene Symbol
H2BFWT
Entrez Gene ID
158983
Gene Name
H2B histone family member W, testis specific
Gene Aliases
Chromosome Location
Chr. X: 104011147 - 104013687 on Build GRCh38
UniGene ID
Hs.127778
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002916.4 NP_001002916.3 3 668
GenBank mRNA BC038109.2 - 3 665
BC118604.1 - 3 661 40109007
BC121816.1 - 3 661 40109006

View Full Product Details