Gene Transcripts

Gene Symbol
MON1B
Entrez Gene ID
22879
Gene Name
MON1 homolog B, secretory trafficking associated
Gene Aliases
HSRG1, SAND2, SRG1
Chromosome Location
Chr. 16: 77190919 - 77199646 on Build GRCh38
UniGene ID
Hs.513743
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014940.3 NP_055755.1 2, 3 499
GenBank mRNA AB020679.1 - 2, 3 183
AF442486.1 - 1, 2 163
AK023374.1 - 2, 3 449
AK023896.1 - 2, 3 227
BC024277.1 - 2, 3 479 4843859
CF125013.1 - 2, 3 456

View Full Product Details