Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000499.4 | NP_000490.1 | 2 | 310 | |
NM_001319216.1 | NP_001306145.1 | 2 | 313 | ||
NM_001319217.1 | NP_001306146.1 | 2 | 313 | ||
XM_017021953.1 | XP_016877442.1 | 2 | 313 | ||
GenBank mRNA | AK223108.1 | - | 2 | 231 | |
AK223113.1 | - | 2 | 228 | ||
AK310810.1 | - | 2 | 357 | ||
AK313880.1 | - | 2 | 228 | ||
AM233519.1 | - | 2 | 191 | ||
AM233520.1 | - | 2 | 191 | ||
AM236046.1 | - | 2 | 191 | ||
AY310359.1 | - | 1 | 114 | ||
BC023019.1 | - | 2 | 195 | 5123393 | |
DA693879.1 | - | 2 | 313 | ||
K03191.1 | - | 2 | 200 |