Gene Transcripts

Gene Symbol
SAMD4A
Entrez Gene ID
23034
Gene Name
sterile alpha motif domain containing 4A
Gene Aliases
SAMD4, SMAUG, SMAUG1, SMG, SMGA
Chromosome Location
Chr. 14: 54567612 - 54793315 on Build GRCh38
UniGene ID
Hs.733324
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161576.2 NP_001155048.2 8, 9 1958
NM_001161577.1 NP_001155049.1 5, 6 851
NM_015589.5 NP_056404.4 9, 10 2222
GenBank mRNA AB028976.1 - 7, 8 1289
AK024652.1 - 9, 10 1915
AK025923.1 - 7, 8 1103
AL117523.1 - 1, 2 172
BC021060.1 - 5, 6 768
BC021726.2 - 5, 6 806
BC121173.1 - 8, 9 1655 40122465
BC121174.1 - 8, 9 1655 40122467

View Full Product Details