Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015885.3 | NP_056969.2 | 5 | 1217 | |
XM_005274048.1 | XP_005274105.1 | 5 | 1217 | ||
XM_005274049.1 | XP_005274106.1 | 5 | 1217 | ||
XM_017017918.1 | XP_016873407.1 | 5 | 1217 | ||
GenBank mRNA | AB020631.1 | - | 5 | 1141 | |
AF046935.1 | - | 5 | 1169 | ||
AK025103.1 | - | 5 | 1153 | ||
AK302606.1 | - | 5 | 1186 | ||
BC065384.1 | - | 5 | 1109 | 6043635 | |
BC146778.1 | - | 5 | 1141 | 9007228 |