Gene Transcripts

Gene Symbol
EIF6
Entrez Gene ID
3692
Gene Name
eukaryotic translation initiation factor 6
Gene Aliases
CAB, EIF3A, ITGB4BP, b(2)gcn, eIF-6, p27(BBP), p27BBP
Chromosome Location
Chr. 20: 35278906 - 35284816 on Build GRCh38
UniGene ID
Hs.743992
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001267810.1 NP_001254739.1 2 47
NM_002212.3 NP_002203.1 2 117
NM_181466.2 NP_852131.1 2 117
NM_181468.2 NP_852133.1 1 251
NR_052022.1 2 117
NR_052023.1 2 117
NR_052024.1 2 47
GenBank mRNA AB062289.1 - 2 50
AF022229.1 - 1 106
AF047433.1 - 2 91
AK296076.1 - 2 63
AK314969.1 - 2 93
AL537456.3 - 1 244
BC001119.2 - 2 90 2989973
BC011845.2 - 2 68 4120463
BC019305.1 - 2 50 2823211
BC095465.1 - 2 66
BG283650.1 - 1 40
BG334381.1 - 71
BM794263.1 - 2 71
BQ222221.1 - 1 37
CN265282.1 - 1 251
CR456764.1 - 1 21
CT002623.1 - 2 69
DB448068.1 - 2 118
Y11435.1 - 2 59

View Full Product Details