Gene Transcripts

Gene Symbol
AMOTL2
Entrez Gene ID
51421
Gene Name
angiomotin like 2
Gene Aliases
LCCP
Chromosome Location
Chr. 3: 134355345 - 134375417 on Build GRCh38
UniGene ID
Hs.426312
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278683.1 NP_001265612.1 6 1739
NM_001278685.1 NP_001265614.1 6 1753
NM_016201.3 NP_057285.3 6 1753
XM_006713654.2 XP_006713717.1 6 1771
XM_011512881.1 XP_011511183.1 6 1610
XM_017006580.1 XP_016862069.1 6 1610
XM_017006581.1 XP_016862070.1 6 1791
XM_017006582.1 XP_016862071.1 6 1618
GenBank mRNA AB023206.1 - 6 1686
AF175966.1 - 5 820
AK001343.1 - 6 1616
AK074731.1 - 1 151
AK091040.1 - 3 821
AK223598.1 - 6 1611
AK291616.1 - 6 1697
AK299270.1 - 6 1739
BC011454.1 - 4 719 4181482
BC025981.2 - 3 309

View Full Product Details