Gene Transcripts
- Gene Symbol
- RRP7A
- Entrez Gene ID
- 27341
- Gene Name
- ribosomal RNA processing 7 homolog A
- Gene Aliases
- BK126B4.3, CGI-96
- Chromosome Location
- Chr. 22: 42508335 - 42519823 on Build GRCh38
- UniGene ID
- Hs.745006
- Species
- Homo sapiens
- Gene Symbol
- RRP7BP
- Entrez Gene ID
- 91695
- Gene Name
- ribosomal RNA processing 7 homolog B, pseudogene
- Gene Aliases
- RRP7B, dJ222E13.2
- Chromosome Location
- Chr. 22: 42573244 - 42582011 on Build GRCh38
- UniGene ID
- Hs.534041
- Species
- Homo sapiens
View Full Product Details