Gene Transcripts
- Gene Symbol
- COA1
- Entrez Gene ID
- 55744
- Gene Name
- cytochrome c oxidase assembly factor 1 homolog
- Gene Aliases
- C7orf44, MITRAC15
- Chromosome Location
- Chr. 7: 43608452 - 43729541 on Build GRCh38
- UniGene ID
- Hs.268887
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001321197.1 |
NP_001308126.1
|
5, 6 |
637 |
|
NM_001321198.1 |
NP_001308127.1
|
5, 6 |
518 |
|
NM_001321199.1 |
NP_001308128.1
|
5, 6 |
482 |
|
NM_001321200.1 |
NP_001308129.1
|
5, 6 |
538 |
|
NM_001321201.1 |
NP_001308130.1
|
4, 5 |
410 |
|
NM_018224.3 |
NP_060694.2
|
4, 5 |
417 |
|
NR_047701.1 |
|
5, 6 |
482 |
|
NR_135580.1 |
|
5, 6 |
518 |
|
NR_135581.1 |
|
5, 6 |
518 |
|
NR_135582.1 |
|
4, 5 |
417 |
|
NR_135583.1 |
|
4, 5 |
417 |
|
XM_005249807.4 |
XP_005249864.1
|
4, 5 |
481 |
|
XM_006715752.2 |
XP_006715815.1
|
5, 6 |
449 |
|
XM_017012413.1 |
XP_016867902.1
|
4, 5 |
408 |
|
XM_017012414.1 |
XP_016867903.1
|
4, 5 |
408 |
|
XM_017012415.1 |
XP_016867904.1
|
5, 6 |
510 |
|
XM_017012416.1 |
XP_016867905.1
|
4, 5 |
408 |
|
XM_017012417.1 |
XP_016867906.1
|
4, 5 |
408 |
|
XM_017012418.1 |
XP_016867907.1
|
4, 5 |
408 |
|
XM_017012419.1 |
XP_016867908.1
|
4, 5 |
408 |
|
XM_017012420.1 |
XP_016867909.1
|
5, 6 |
510 |
|
XM_017012421.1 |
XP_016867910.1
|
5, 6 |
632 |
|
XM_017012422.1 |
XP_016867911.1
|
5, 6 |
521 |
|
XM_017012423.1 |
XP_016867912.1
|
5, 6 |
441 |
|
XM_017012424.1 |
XP_016867913.1
|
4, 5 |
424 |
|
XM_017012425.1 |
XP_016867914.1
|
5, 6 |
525 |
|
XM_017012426.1 |
XP_016867915.1
|
5, 6 |
644 |
|
XM_017012427.1 |
XP_016867916.1
|
6, 7 |
599 |
|
XM_017012428.1 |
XP_016867917.1
|
4, 5 |
424 |
|
XM_017012429.1 |
XP_016867918.1
|
3, 4 |
318 |
|
XM_017012430.1 |
XP_016867919.1
|
5, 6 |
510 |
|
XM_017012431.1 |
XP_016867920.1
|
4, 5 |
424 |
|
XM_017012432.1 |
XP_016867921.1
|
4, 5 |
424 |
|
GenBank mRNA |
AK001665.1 |
- |
5, 6 |
451 |
|
AK021905.1 |
- |
5, 6 |
587 |
|
AK023007.1 |
- |
5, 6 |
434 |
|
AK023053.1 |
- |
5, 6 |
450 |
|
AK097203.1 |
- |
3, 4 |
1948 |
|
AK293043.1 |
- |
5, 6 |
460 |
|
BC056884.1 |
- |
5, 6 |
409 |
6159045 |
BG028456.1 |
- |
|
439 |
|
BG334368.1 |
- |
5, 6 |
427 |
|
BG548154.1 |
- |
5, 6 |
474 |
|
BM918741.1 |
- |
4, 5 |
390 |
|
BQ231567.1 |
- |
4, 5 |
355 |
|
DA742258.1 |
- |
4, 5 |
399 |
|
View Full Product Details