Gene Transcripts

Gene Symbol
DNAJB13
Entrez Gene ID
374407
Gene Name
DnaJ heat shock protein family (Hsp40) member B13
Gene Aliases
RSPH16A, TSARG5, TSARG6
Chromosome Location
Chr. 11: 73950319 - 73970287 on Build GRCh38
UniGene ID
Hs.567888
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153614.3 NP_705842.2 6 1433
XM_005273984.3 XP_005274041.1 5 798
XM_011545004.2 XP_011543306.1 8 1684
XM_011545005.2 XP_011543307.1 7 1524
XM_011545007.2 XP_011543309.1 6 1209
XM_011545009.2 XP_011543311.1 8 1358
XM_011545015.2 XP_011543317.1 3 551
XM_017017675.1 XP_016873164.1 7 954
XM_017017676.1 XP_016873165.1 8 1534
XM_017017677.1 XP_016873166.1 7 1403
XM_017017678.1 XP_016873167.1 6 1214
GenBank mRNA AF516185.1 - 3 1405
AK058095.1 - 3 1405
AK128589.1 - 8 3478
AY138810.2 - 6 1433
AY325766.1 - 6 726
BC127689.1 - 1 540

View Full Product Details