Gene Transcripts

Gene Symbol
GRK1
Entrez Gene ID
6011
Gene Name
G protein-coupled receptor kinase 1
Gene Aliases
GPRK1, RHOK, RK
Chromosome Location
Chr. 13: 113645771 - 113735664 on Build GRCh38
UniGene ID
Hs.103501
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002929.2 NP_002920.1 5 1258
XM_017020684.1 XP_016876173.1 5 2925
GenBank mRNA AK289912.1 - 5 1382
BC140945.1 - 5 1223 9056757
L77503.1 - 5 1165
U63973.1 - 5 1258

View Full Product Details