Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016433.3 | NP_057517.1 | 2 | 260 | |
XM_011538434.2 | XP_011536736.1 | 2 | 213 | ||
GenBank mRNA | AF209704.1 | - | 2 | 156 | |
AK313457.1 | - | 2 | 149 | ||
AY372530.1 | - | 2 | 146 | ||
AY372531.1 | - | 2 | 146 | ||
AY372532.1 | - | 2 | 175 | ||
BC009932.2 | - | 2 | 260 | 3959556 | |
JN039379.1 | - | 2 | 146 |