Gene Transcripts

Gene Symbol
EMC4
Entrez Gene ID
51234
Gene Name
ER membrane protein complex subunit 4
Gene Aliases
PIG17, TMEM85
Chromosome Location
Chr. 15: 34224997 - 34230165 on Build GRCh38
UniGene ID
Hs.250905
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286420.1 NP_001273349.1 3 397
NM_016454.3 NP_057538.1 3 397
GenBank mRNA AF151018.1 - 3 350
AK075227.1 - 3 376
AK290524.1 - 3 381
AY336092.1 - 3 337
BC002583.2 - 3 362 3161718
BG707804.1 - 3 410
KU178640.1 - 3 279
KU178641.1 - 3 279

View Full Product Details