Gene Transcripts

Gene Symbol
NKIRAS2
Entrez Gene ID
28511
Gene Name
NFKB inhibitor interacting Ras like 2
Gene Aliases
KBRAS2, kappaB-Ras2
Chromosome Location
Chr. 17: 42017576 - 42025641 on Build GRCh38
UniGene ID
Hs.632252
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001349.2 NP_001001349.1 3 379
NM_001144927.1 NP_001138399.1 3 483
NM_001144929.1 NP_001138401.1 4 367
NM_017595.5 NP_060065.2 3 464
XM_005257251.1 XP_005257308.1 3 491
GenBank mRNA AF229840.1 - 2 314
AK027265.1 - 3 455
AK054571.1 - 3 491
AL137682.1 - 1 2204
BC007316.2 - 3 416
BC007450.2 - 3 372 3028708
BC063498.1 - 3 935 5248950
BG717103.1 - 3 485
DA509824.1 - 4 336
DA607452.1 - 3 462

View Full Product Details